1,1-Cyclobutanedicarboxylic acid
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
Cyclobutane-1,1-dicarboxylic acid
003367-25G
003367-100G
FLUO003367-25GEA
0
GBP
FLUO003367-25G
FLUO003367-100G
1,1-Cyclobutanedicarboxylic acid
1,1-Cyclobutanedicarboxylic acid
Formula:
C₆H₈O₄ MW: 144.13 g/mol Boiling Pt: 663 °C (1013 hPa) Melting Pt: 156…158 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001325 CAS Number: 5445-51-2 EINECS: 226-651-2 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...