Cyclopentanecarboxylic acid

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

ARC-1454 ARC-1454-1
BTRNARC-1454EA 0 GBP
BTRNARC-1454 BTRNARC-1454-1
Cyclopentanecarboxylic acid
Cyclopentanecarboxylic acid
Formula: C₆H₁₀O₂
MW: 114.14 g/mol
Boiling Pt: 216 °C (1013 hPa)
Melting Pt: 4 °C
Density: 1.05 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001371
CAS Number: 3400-45-1
EINECS: 222-269-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR