6-Methoxyquinaldine
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
002592-1G
002592-5G
FLUO002592-1GEA
0
GBP
FLUO002592-1G
FLUO002592-5G
6-Methoxyquinaldine
6-Methoxyquinaldine
Formula:
C₁₁H₁₁NO Boiling Pt: 95…96 °C (0.5 mmHg) Melting Pt: 67 °C |
MDL Number:
MFCD00006761 CAS Number: 1078-28-0 EINECS: 214-080-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...