6-Fluoroquinaldine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

002573-5G 002573-1G
FLUO002573-5GEA 0 GBP
FLUO002573-5G FLUO002573-1G
6-Fluoroquinaldine
6-Fluoroquinaldine
Formula: C₁₀H₈FN
MDL Number: MFCD00041233
CAS Number: 1128-61-6
EINECS: 214-439-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR