8-Bromoquinoline ≥98%

Supplier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

H25961.MD H25961.03
H25961.MDEA 26.2 GBP
H25961.MD H25961.03
8-Bromoquinoline ≥98%
8-Bromoquinoline

Purity: 98% CAS No.: 16567-18-3 Molecular Formula: C9H6BrN Formula Weight: 208.06 Flash Point: 110°(230°F) Refractive Index: 1.6725 Boiling Point: 302-304° MDL No.: MFCD00191859

Formula: C₉H₆BrN
MW: 208.06 g/mol
Boiling Pt: 302…304 °C
Density: 1.594
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00191859
CAS Number: 16567-18-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR