1,3,5-Triphenylbenzene, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M-Clarity™ quality level = MQ100
T82007-5G
T82007-100G
SIALT82007-5GEA
51
GBP
SIALT82007-5G
SIALT82007-100G
1,3,5-Triphenylbenzene, Sigma-Aldrich®
1,3,5-Triphenylbenzene
Formula:
C₂₄H₁₈ MW: 306.41 g/mol Boiling Pt: 460 °C (1013 hPa) Melting Pt: 176 °C Density: 1.22 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003060 CAS Number: 612-71-5 EINECS: 210-318-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...