1,3,5-Triisopropylbenzene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

022115-25G 022115-100G
FLUO022115-25GEA 0 GBP
FLUO022115-25G FLUO022115-100G
1,3,5-Triisopropylbenzene
1,3,5-Triisopropylbenzene
Formula: C₁₅H₂₄
Boiling Pt: 232 °C (1013 hPa)
Melting Pt: –7 °C
Density: 0.8447 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008890
CAS Number: 717-74-8
EINECS: 211-941-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR