4-Chloromethyl-o-xylene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

018620-10G 018620-25G
FLUO018620-10GEA 0 GBP
FLUO018620-10G FLUO018620-25G
4-Chloromethyl-o-xylene
4-Chloromethyl-o-xylene
Formula: C₉H₁₁Cl
MDL Number: MFCD00000910
CAS Number: 102-46-5
EINECS: 203-032-5
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR