3,5-Dimethylthiophenol
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
1,3-Dimethyl-5-mercaptobenzene, 3,5-Dimethylbenzene-1-thiol, 3,5-Dimethylbenzenethiol, 5-Mercapto-m-xylene
001348-5G
001348-25G
FLUO001348-5GEA
0
GBP
FLUO001348-5G
FLUO001348-25G
3,5-Dimethylthiophenol
3,5-Dimethylthiophenol
Formula:
C₈H₁₀S MW: 138.23 g/mol Boiling Pt: 96 °C (1013 hPa) Density: 1.02 g/cm³ (20 °C) |
MDL Number:
MFCD00040226 CAS Number: 38360-81-5 EINECS: 253-901-8 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...