4-Iodo-m-xylene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

001771-25G 001771-100G
FLUO001771-25GEA 0 GBP
FLUO001771-25G FLUO001771-100G
4-Iodo-m-xylene
4-Iodo-m-xylene
Formula: C₈H₉I
Boiling Pt: 112 °C (1013 hPa)
Density: 1,6609 g/cm³ (13 °C)
MDL Number: MFCD00013706
CAS Number: 4214-28-2
EINECS: 224-141-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR