2-Fluoro-m-xylene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Fluoro-1,3-dimethylbenzene, 1-Fluoro-2,6-dimethylbenzene

003242-5G 003242-25G
FLUO003242-5GEA 0 GBP
FLUO003242-5G FLUO003242-25G
2-Fluoro-m-xylene
2-Fluoro-m-xylene
Formula: C₈H₉F
MW: 124.16 g/mol
Boiling Pt: 165…167 °C (1013 hPa)
Density: 0.988 g/cm³ (20 °C)
MDL Number: MFCD00039215
CAS Number: 443-88-9
EINECS: 207-144-5
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR