α,α,α,α',α',α'-Hexafluoro-m-xylene
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
1,3-Bis(trifluoromethyl)benzene, α,α,α,β,β,β-Hexafluoro-m-xylene
001600-1KG
001600-250G
FLUO001600-1KGEA
0
GBP
FLUO001600-1KG
FLUO001600-250G
α,α,α,α',α',α'-Hexafluoro-m-xylene
α,α,α,α',α',α'-Hexafluoro-m-xylene
Formula:
C₈H₄F₆ MW: 214.11 g/mol Boiling Pt: 116.3 °C (1013 hPa) Melting Pt: –35 °C Density: 1.383 g/cm³ (22 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00000392 CAS Number: 402-31-3 EINECS: 206-939-4 UN: 1993 ADR: 3,III |
Learn more
![](/m-uk.vwr.com/en_GB/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...