4-Chloro-o-xylene
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
018461-5G
018461-25G
FLUO018461-5GEA
0
GBP
FLUO018461-5G
FLUO018461-25G
4-Chloro-o-xylene
4-Chloro-o-xylene
Formula:
C₈H₉Cl Boiling Pt: 194…195 °C (1013 hPa) Melting Pt: –6 °C Density: 1,0682 g/cm³ (15 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00000596 CAS Number: 615-60-1 EINECS: 210-438-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...