Paraldehyde, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
76260-1L
76260-250ML
SIAL76260-1LEA
121
GBP
SIAL76260-1L
SIAL76260-250ML
Paraldehyde, Sigma-Aldrich®
Paraldehyde
Formula:
C₆H₁₂O₃ |
MDL Number:
MFCD00036208 CAS Number: 123-63-7 EINECS: 204-639-8 UN: 1264 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...