(R)-(+)-α-Methylbenzylamine, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
(R)-(+)-α-Phenylethylamine, (R)-(+)-1-Phenylethylamine, D(+)-α-Methylbenzylamine
726621-100G
726621-500G
SIAL726621-100GEA
85
GBP
SIAL726621-100G
SIAL726621-500G
(R)-(+)-α-Methylbenzylamine, Sigma-Aldrich®
(R)-(+)-α-Methylbenzylamine
Formula:
C₈H₁₁N MW: 121.18 g/mol Boiling Pt: 180 °C (1013 hPa) Melting Pt: 32.5 °C Density: 0.95 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00064405 CAS Number: 3886-69-9 EINECS: 223-423-4 Merck Index: 12,06106 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...