N-Methylmorpholine N-oxide monohydrate ≥95%, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
4-Methylmorpholine N-oxide monohydrate, 4-Methylmorpholine 4-oxide hydrate, NMMO
67873-100G
67873-25G
SIAL67873-100GEA
246
GBP
SIAL67873-100G
SIAL67873-25G
N-Methylmorpholine N-oxide monohydrate ≥95%, Sigma-Aldrich®
N-Methylmorpholine N-oxide monohydrate
Formula:
C₅H₁₁NO₂·1H₂O MW: 135.16 g/mol Melting Pt: 71-75 °C |
MDL Number:
MFCD00149388 CAS Number: 70187-32-5 EINECS: 231-391-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...