4-(4-Bromophenyl)morpholine, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ100

591777-25G 591777-5G
SIAL591777-25GEA 238 GBP
SIAL591777-25G SIAL591777-5G
4-(4-Bromophenyl)morpholine, Sigma-Aldrich®
4-(4-Bromophenyl)morpholine
Formula: C₁₀H₁₂BrNO
MW: 242.12 g/mol
MDL Number: MFCD04112483
CAS Number: 30483-75-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR