1,3-Diphenylurea, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: N,N’-Diphenylurea, N,N'-Diphenylurea

M-Clarity™ quality level = MQ100

142158-100G 142158-25G
SIAL142158-100GEA 149 GBP
SIAL142158-100G SIAL142158-25G
1,3-Diphenylurea, Sigma-Aldrich®
1,3-Diphenylurea
Formula: C₁₃H₁₂N₂O
MDL Number: MFCD00003017
CAS Number: 102-07-8
EINECS: 203-003-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR