2-Bromo-6-methoxynaphthalene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

023276-100G 023276-25G
FLUO023276-100GEA 0 GBP
FLUO023276-100G FLUO023276-25G
2-Bromo-6-methoxynaphthalene
2-Bromo-6-methoxynaphthalene
Formula: C₁₁H₉BrO
MDL Number: MFCD00004062
CAS Number: 5111-65-9
EINECS: 225-837-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR