4-Bromothiophene-2-carbaldehyde

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-Bromo-2-thiophenecarboxaldehyde, 4-Bromothiophene-2-carboxaldehyde

019228-10G 019228-25G
FLUO019228-10GEA 0 GBP
FLUO019228-10G FLUO019228-25G
4-Bromothiophene-2-carbaldehyde
4-Bromothiophene-2-carbaldehyde
Formula: C₅H₃BrOS
MW: 191.05 g/mol
MDL Number: MFCD00005431
CAS Number: 18791-75-8
EINECS: 242-577-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR