1-Isopropyl-2-thiourea
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
018760-25G
018760-5G
FLUO018760-25GEA
0
GBP
FLUO018760-25G
FLUO018760-5G
1-Isopropyl-2-thiourea
1-Isopropyl-2-thiourea
Formula:
C₄H₁₀N₂S |
MDL Number:
MFCD00041375 CAS Number: 1719-76-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...