3-Methylthiophene-2-carbonyl chloride
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
FL14753.1
014753-5G
014753-25G
CHMPFL14753.1EA
0
GBP
CHMPFL14753.1
FLUO014753-5G
FLUO014753-25G
3-Methylthiophene-2-carbonyl chloride
3-Methylthiophene-2-carbonyl chloride
Formula:
C₆H₅ClOS |
MDL Number:
MFCD00130091 CAS Number: 61341-26-2 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...