2-Chlorophenethylamine
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
008893-5G
008893-25G
FLUO008893-5GEA
0
GBP
FLUO008893-5G
FLUO008893-25G
2-Chlorophenethylamine
2-Chlorophenethylamine
Formula:
C₈H₁₀ClN MW: 155.63 g/mol |
MDL Number:
MFCD00008185 CAS Number: 13078-80-3 EINECS: 235-982-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...