4-(Trifluoromethoxy)benzamidoxime

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

007538-1G 007538-5G
FLUO007538-1GEA 0 GBP
FLUO007538-1G FLUO007538-5G
4-(Trifluoromethoxy)benzamidoxime
4-(Trifluoromethoxy)benzamidoxime
Formula: C₈H₇F₃N₂O₂
MDL Number: MFCD00085175
CAS Number: 56935-71-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR