2-(Dimethylamino)-6-fluorobenzonitrile

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

004927-1G 004927-5G
FLUO004927-1GEA 0 GBP
FLUO004927-1G FLUO004927-5G
2-(Dimethylamino)-6-fluorobenzonitrile
2-(Dimethylamino)-6-fluorobenzonitrile
Formula: C₉H₉FN₂
MDL Number: MFCD00052221
CAS Number: 96994-73-9
UN: 3276
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR