Diethyl hexafluoroglutarate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

004765-1G 004765-5G
FLUO004765-1GEA 0 GBP
FLUO004765-1G FLUO004765-5G
Diethyl hexafluoroglutarate
Diethyl hexafluoroglutarate
Formula: C₉H₁₀F₆O₄
MDL Number: MFCD00015156
CAS Number: 424-40-8
EINECS: 207-035-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR