Perfluoro(methylcyclohexane)
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
FlutecR PP2
003295-100G
003295-500G
FLUO003295-100GEA
134
GBP
FLUO003295-100G
FLUO003295-500G
Perfluoro(methylcyclohexane)
Perfluoro(methylcyclohexane)
Formula:
C₇F₁₄ MW: 350.05 g/mol |
MDL Number:
MFCD00001468 CAS Number: 355-02-2 EINECS: 206-573-5 |
Learn more
![](/m-uk.vwr.com/en_GB/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...