Decafluoro-4,6-bis(trifluoromethyl)cyclohexane
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
Perfluoro-1,3-dimethylcyclohexane, Perfluoro(1,3-dimethylcyclohexane)
003287-25G
003287-100G
003287-500G
FLUO003287-25GEA
0
GBP
FLUO003287-25G
FLUO003287-100G
FLUO003287-500G
Decafluoro-4,6-bis(trifluoromethyl)cyclohexane
Decafluoro-4,6-bis(trifluoromethyl)cyclohexane
Formula:
C₈F₁₆ |
MDL Number:
MFCD00001469 CAS Number: 335-27-3 EINECS: 206-386-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...