Dimethyl hexafluoroglutarate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Dimethyl perfluoroglutarate

002289-5G 002289-25G
FLUO002289-5GEA 0 GBP
FLUO002289-5G FLUO002289-25G
Dimethyl hexafluoroglutarate
Dimethyl hexafluoroglutarate
Formula: C₇H₆F₆O₄
MDL Number: MFCD00155851
CAS Number: 1513-62-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR