4-Butoxyaniline

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

001735-5G 001735-25G
FLUO001735-5GEA 0 GBP
FLUO001735-5G FLUO001735-25G
4-Butoxyaniline
4-Butoxyaniline
Formula: C₁₀H₁₅NO
MDL Number: MFCD00007866
CAS Number: 4344-55-2
EINECS: 224-402-2
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR