4-Bromo-2-thenoic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-Bromo-2-thiophenecarboxylic acid

HI-1319-25G HI-1319-5G HI-1319-1G HI-1319-100G
COBBHI-1319-25GEA 0 GBP
COBBHI-1319-25G COBBHI-1319-5G COBBHI-1319-1G COBBHI-1319-100G
4-Bromo-2-thenoic acid
4-Bromo-2-thenoic acid
Formula: C₅H₃BrO₂S
MW: 207.05 g/mol
MDL Number: MFCD03422294
CAS Number: 16694-18-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR