3-(Ethylthio)phenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BB-8367-5G BB-8367-1G
COBBBB-8367-5GEA 0 GBP
COBBBB-8367-5G COBBBB-8367-1G
3-(Ethylthio)phenylboronic acid
3-(Ethylthio)phenylboronic acid
Formula: C₈H₁₁BO₂S
MW: 182.05 g/mol
MDL Number: MFCD03092930
CAS Number: 870718-05-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR