4-(4-Bromophenyl)morpholine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AN-1437-5G AN-1437-25G AN-1437-1G
COBBAN-1437-5GEA 0 GBP
COBBAN-1437-5G COBBAN-1437-25G COBBAN-1437-1G
4-(4-Bromophenyl)morpholine
4-(4-Bromophenyl)morpholine
Formula: C₁₀H₁₂BrNO
MW: 242.12 g/mol
MDL Number: MFCD04112483
CAS Number: 30483-75-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR