L(+)-Citrulline

Supplier: BACHEM BIOCHEMICA
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: H-Cit-OH

4031894.0100 4031894.0250
BACE4031894.0100EA 0 GBP
BACE4031894.0100 BACE4031894.0250
L(+)-Citrulline
L(+)-Citrulline
Formula: C₆H₁₃N₃O₃
MDL Number: MFCD00064397
CAS Number: 372-75-8
EINECS: 206-759-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR