Fmoc-D-Pro-OH
Supplier: BACHEM BIOCHEMICA
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
N-Fmoc-D-proline
4011894.0005
4011894.0025
BACE4011894.0005EA
0
GBP
BACE4011894.0005
BACE4011894.0025
Fmoc-D-Pro-OH
Fmoc-D-Pro-OH
Formula:
C₂₀H₁₉NO₄ |
MDL Number:
MFCD00077067 CAS Number: 101555-62-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...