Fmoc-Pro-OH

Supplier: BACHEM BIOCHEMICA
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-proline, N-Fmoc-L-proline

4003150.0025 4003150.0100 4003150.0500
BACE4003150.0025EA 0 GBP
BACE4003150.0025 BACE4003150.0100 BACE4003150.0500
Fmoc-Pro-OH
Fmoc-Pro-OH
Formula: C₂₀H₁₉NO₄
MW: 337.38 g/mol
MDL Number: MFCD00037122
CAS Number: 71989-31-6
EINECS: 276-259-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR