3-Bromothiophene-2-carbaldehyde ≥94%
Supplier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
3-Bromothiophene-2-carboxaldehyde, 3-Bromo-2-formylthiophene
L19386.03
L19386.06
L19386.03EA
0
GBP
L19386.03
L19386.06
3-Bromothiophene-2-carbaldehyde ≥94%
3-Bromothiophene-2-carbaldehyde
Purity: 94%
CAS No.: 930-96-1
Molecular Formula: C5H3BrOS
Formula Weight: 191.05
Refractive Index: 1.6377
Boiling Point: 110-113°/10mm
MDL No.: MFCD00126680
Formula:
C₅H₃BrOS Boiling Pt: 110…113 °C (10 mmHg) Storage Temperature: Ambient |
MDL Number:
MFCD00126680 CAS Number: 930-96-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...