2-Bromo-3,3,3-trifluoropropene ≥97%
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
2-Bromo-3,3,3-trifluoro-1-propene
B24614.14
B24614.06
B24614.14EA
182
GBP
B24614.14
B24614.06
2-Bromo-3,3,3-trifluoropropene ≥97%
2-Bromo-3,3,3-trifluoropropene
Purity: 97%
CAS No.: 1514-82-5
Molecular Formula: C3H2BrF3
Formula Weight: 174.95
Refractive Index: 1.3530
Formula:
C₃H₂BrF₃ MW: 174.95 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00077469 CAS Number: 1514-82-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...