1-Propyl-2-urea ≥98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

A10160.06 A10160.14
A10160.06EA 25.6 GBP
A10160.06 A10160.14
1-Propyl-2-urea ≥98%
1-Propyl-2-urea

Purity: 98% CAS No.: 627-06-5 Molecular Formula: C4H10N2O Formula Weight: 102.14 Melting Point: 99-104° MDL No.: MFCD00014793

Formula: C₄H₁₀N₂O
Melting Pt: 99…104 °C
Storage Temperature: Ambient
MDL Number: MFCD00014793
CAS Number: 627-06-5
EINECS: 210-981-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR