α'-Bromo-α,α,α-trifluoro-p-xylene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: 4-(Bromomethyl)benzotrifluoride, 1-(Bromomethyl)-4-(trifluoromethyl)benzene, 4-(Trifluoromethyl)benzyl bromide

007607-5G 007607-25G 007607-100G
FLUO007607-5GEA 0 GBP
FLUO007607-5G FLUO007607-25G FLUO007607-100G
α'-Bromo-α,α,α-trifluoro-p-xylene
α'-Bromo-α,α,α-trifluoro-p-xylene
Formula: C₈H₆BrF₃
MW: 239.03 g/mol
Boiling Pt: 73…75 °C (0.4 mmHg)
Melting Pt: 31.5
Density: 1.54 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000403
CAS Number: 402-49-3
EINECS: 206-947-8
UN: 3261
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR