α'-Bromo-α,α,α-trifluoro-m-xylene
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
3-(Trifluoromethyl)benzyl bromide, 1-(Bromomethyl)-3-(trifluoromethyl)benzene, 3-(Bromomethyl)benzotrifluoride
007606-5G
007606-25G
007606-100G
FLUO007606-5GEA
0
GBP
FLUO007606-5G
FLUO007606-25G
FLUO007606-100G
α'-Bromo-α,α,α-trifluoro-m-xylene
α'-Bromo-α,α,α-trifluoro-m-xylene
Formula:
C₈H₆BrF₃ Boiling Pt: 50 °C (0.3 mmHg) Density: 1.56 g/cm³ (20 °C) |
MDL Number:
MFCD00000395 CAS Number: 402-23-3 EINECS: 206-938-9 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...