α'-Bromo-α,α,α-trifluoro-o-xylene
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
2-(Trifluoromethyl)benzyl bromide, 1-(Bromomethyl)-2-(trifluoromethyl)benzene, 2-(Bromomethyl)benzotrifluoride
007605-5G
007605-25G
007605-100G
FLUO007605-5GEA
0
GBP
FLUO007605-5G
FLUO007605-25G
FLUO007605-100G
α'-Bromo-α,α,α-trifluoro-o-xylene
α'-Bromo-α,α,α-trifluoro-o-xylene
Formula:
C₈H₆BrF₃ Boiling Pt: 116…118 °C (60 mmHg) Melting Pt: 30 °C Density: 1.57 g/cm³ (20 °C) |
MDL Number:
MFCD00013558 CAS Number: 395-44-8 EINECS: 206-901-7 UN: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...