4-Bromo-o-xylene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

018328-100G 018328-25G
FLUO018328-100GEA 0 GBP
FLUO018328-100G FLUO018328-25G
4-Bromo-o-xylene
4-Bromo-o-xylene
Formula: C₈H₉Br
Boiling Pt: 215 °C (1013 hPa)
Density: 1.37 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000071
CAS Number: 583-71-1
EINECS: 209-518-3
UN: 1701
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR