2,5-Dibromo-p-xylene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2,5-Dibromo-1,4-dimethylbenzene, 1,4-Dibromo-2,5-dimethylbenzene

019177-250G 019177-50G
FLUO019177-250GEA 0 GBP
FLUO019177-250G FLUO019177-50G
2,5-Dibromo-p-xylene
2,5-Dibromo-p-xylene
Formula: C₈H₈Br₂
MW: 263.96 g/mol
Boiling Pt: 261 °C (1013 hPa)
Melting Pt: 75 °C
MDL Number: MFCD00000091
CAS Number: 1074-24-4
EINECS: 214-038-2
UN: 3077
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR