4-Methylveratrole

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 1,2-Dimethoxy-4-methylbenzene, 3,4-Dimethoxytoluene

018601-5G 018601-25G
FLUO018601-5GEA 0 GBP
FLUO018601-5G FLUO018601-25G
4-Methylveratrole
4-Methylveratrole
Formula: C₉H₁₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00016651
CAS Number: 494-99-5
EINECS: 207-796-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR