2-Iodotoluene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 1-Iodo-2-methylbenzene

001199-100G 001199-25G
FLUO001199-100GEA 0 GBP
FLUO001199-100G FLUO001199-25G
2-Iodotoluene
2-Iodotoluene
Formula: C₇H₇I
Boiling Pt: 211 °C (1013 hPa)
Density: 1,71 g/cm³ (20 °C)
Flash Pt: 90 °C
Storage Temperature: Ambient
MDL Number: MFCD00001042
CAS Number: 615-37-2
EINECS: 210-422-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR