3-(Trifluoromethyl)anisole

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

007579-25G 007579-100G
FLUO007579-25GEA 0 GBP
FLUO007579-25G FLUO007579-100G
3-(Trifluoromethyl)anisole
3-(Trifluoromethyl)anisole
Formula: C₈H₇F₃O
MW: 176.14 g/mol
Boiling Pt: 159.5
Melting Pt: –65 °C
Density: 1.2426 g/cm³ (20 °C)
MDL Number: MFCD00000385
CAS Number: 454-90-0
EINECS: 207-229-7
UN: 1993
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR