2-Fluoro-5-(trifluoromethyl)anisole

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

005811-5G 005811-10G 005811-1G
FLUO005811-5GEA 0 GBP
FLUO005811-5G FLUO005811-10G FLUO005811-1G
2-Fluoro-5-(trifluoromethyl)anisole
2-Fluoro-5-(trifluoromethyl)anisole
Formula: C₈H₆F₄O
MW: 194.13 g/mol
Boiling Pt: 151
Melting Pt: –15 °C
MDL Number: MFCD01631564
CAS Number: 261951-78-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR