α,α,α-Trifluoro-3,5-dinitrotoluene
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
3,5-Dinitrobenzotrifluoride
004963-25G
004963-100G
004963-500G
FLUO004963-25GEA
0
GBP
FLUO004963-25G
FLUO004963-100G
FLUO004963-500G
α,α,α-Trifluoro-3,5-dinitrotoluene
α,α,α-Trifluoro-3,5-dinitrotoluene
Formula:
C₇H₃F₃N₂O₄ Boiling Pt: 127…129 °C (9 mmHg) Melting Pt: 50 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007233 CAS Number: 401-99-0 EINECS: 206-935-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...