2-Fluoro-5-nitrobenzotrifluoride
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
2-Fluoro-5-nitrobenzotrifluoride, 1-Fluoro-4-nitro-2-(trifluoromethyl)benzene
CFF624.5
006299-25G
006299-100G
CHMPCFF624.5EA
0
GBP
CHMPCFF624.5
FLUO006299-25G
FLUO006299-100G
2-Fluoro-5-nitrobenzotrifluoride
2-Fluoro-5-nitrobenzotrifluoride
Formula:
C₇H₃F₄NO₂ MW: 209.1 g/mol Boiling Pt: 99…110 °C (35 mmHg) Melting Pt: 22…24 °C Density: 1.52 g/cm³ (20 °C) |
MDL Number:
MFCD00024576 CAS Number: 400-74-8 EINECS: 206-925-8 UN: 2810 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...